Thermo Scientific Chemicals

Safranine O, 85%

Catalog number: 419210250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Safranine O, 85%

Catalog number: 419210250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 419210250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
477-73-6
IUPAC Name
2,7-diamino-1,8-dimethyl-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Molecular Formula
C20H19ClN4
InChI Key
QRYAEWIQIBAZOJ-UHFFFAOYSA-N
SMILES
[Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1
Absorption Ratio P-15/P+15
1.1 to 1.32
BSC Certification
Passes
Appearance (Color)
Dark green to dark brown
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text