Saponina, de Quillaja Saponaria Molina, práct.
Saponina, de Quillaja Saponaria Molina, práct.
Saponina, de Quillaja Saponaria Molina, práct.
Saponina, de Quillaja Saponaria Molina, práct.
Thermo Scientific Chemicals

Saponina, de Quillaja Saponaria Molina, práct.

CAS: 74499-23-3
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
100 g
500 g
2.5 kg
Envase:
Ver todo
Frasco de vidrio
Número de catálogo 419235000
Precio (EUR)
241,40
Precio exclusivo en nuestra web
291,00
Ahorro 49,60 (17%)
Each
-
Añadir al carro de la compra
Cantidad:
500 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
241,40
Precio exclusivo en nuestra web
291,00
Ahorro 49,60 (17%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS74499-23-3
EspecificacionesSpecification SheetHoja de especificaciones
Infrared spectrumConforms
pH3.8 to 4.6 (20 % solution)
Solubility(20 % in water) Clear to hazy foaming solution
Appearance (Form)Powder
Appearance (Color)Dark red to tan
Ver más
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documentos y descargas

Certificados

Hojas de datos de seguridad

Compartir número de catálogo, nombre o enlace.