Sebaconitrile, 98%, Thermo Scientific Chemicals
Sebaconitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sebaconitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 419250250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namedecanedinitrile
Molecular FormulaC10H16N2
InChI KeyDFJYZCUIKPGCSG-UHFFFAOYSA-N
SMILESN#CCCCCCCCCC#N
Molecular Weight (g/mol)164.25
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4470 to 1.4490 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only