Silica gel, for chromatography, 0.030-0.200 mm, 60 A, Thermo Scientific Chemicals
Silica gel, for chromatography, 0.030-0.200 mm, 60 A, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silica gel, for chromatography, 0.030-0.200 mm, 60 A, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Glass Bottle
Catalog number 419292500
Price (USD)/ Each
66.80
-
Add to cart
Quantity:
250 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
66.80
Add to cart
Silica gel, for chromatography, 0.030-0.200 mm, 60 A, Thermo Scientific Chemicals
Catalog number419292500
Price (USD)/ Each
66.80
-
Add to cart
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Loss on drying=<6.0 % (150°C)
Pore volume0.7 to 0.85 ml/g (N2)
Surface area480 to 550 m²/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only