Thermo Scientific Chemicals

Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals

Catalog number: 419330100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals

Catalog number: 419330100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 419330100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1470-61-7
IUPAC Name
silver(1+) (diethylcarbamothioyl)sulfanide
Molecular Formula
C5H10AgNS2
InChI Key
NSVHDIYWJVLAGH-UHFFFAOYSA-M
SMILES
[Ag+].CCN(CC)C([S-])=S
Appearance (Color)
Yellow to green to khaki
Appearance (Form)
Powder
Infrared spectrum
Conforms
Solubility
(50 mg/10 ml Pyridine) Clear bright yellow solution
Suitability
>=0.10 Absorbance (for determination of Arsenic)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text