Thermo Scientific Chemicals

Methanesulfonic acid, silver salt, 99%, Thermo Scientific Chemicals

Catalog number: 419340050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methanesulfonic acid, silver salt, 99%, Thermo Scientific Chemicals

Catalog number: 419340050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 419340050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2386-52-9
IUPAC Name
methanesulfonic acid silver
Molecular Formula
CH4AgO3S
InChI Key
GTBQRHOYAUGRPV-UHFFFAOYSA-N
SMILES
[Ag].CS(O)(=O)=O
Infrared spectrum
Conforms
Titration Argentometric
>=98.5 %
Appearance (Color)
White to beige to light grey
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text