Thermo Scientific Chemicals

Silver nitrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 419350025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Silver nitrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 419350025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 419350025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7761-88-8
IUPAC Name
silver(1+) nitrate
Molecular Formula
AgNO3
InChI Key
SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES
[Ag+].[O-][N+]([O-])=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals
Titration
>=99.0 %
Clarity of solution
Passes test
Free acid
Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text