Silver nitrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Silver nitrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver nitrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 419352500
Price (USD)/ Each
829.65
Online exclusive
922.00 
Save 92.35 (10%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
829.65
Online exclusive
922.00 
Save 92.35 (10%)
Add to cart
Silver nitrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Catalog number419352500
Price (USD)/ Each
829.65
Online exclusive
922.00 
Save 92.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7761-88-8
IUPAC Namesilver(1+) nitrate
Molecular FormulaAgNO3
InChI KeySQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES[Ag+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Titration>=99.0 %
Clarity of solutionPasses test
Free acidPasses test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only