Silver nitrate, 99.5%, for analysis
Silver nitrate, 99.5%, for analysis
Silver nitrate, 99.5%, for analysis
Silver nitrate, 99.5%, for analysis
Thermo Scientific Chemicals

Silver nitrate, 99.5%, for analysis

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 419361000
Price (USD)
452.65
Online Exclusive
503.00
Save 50.35 (10%)
Each
In stock
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
452.65
Online Exclusive
503.00
Save 50.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7761-88-8
IUPAC Namesilver(1+) nitrate
Molecular FormulaAgNO3
InChI KeySQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES[Ag+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Cadmium (Cd)=<1 ppm
Nickel (Ni)=<0.2 ppm
Palladium (Pd)=<0.1 ppm
Zinc (Zn)=<10 ppm
Chloride (Cl)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection