Sodium bisulfate monohydrate, 97%, extra pure, reagent, crystals, Thermo Scientific Chemicals
Sodium bisulfate monohydrate, 97%, extra pure, reagent, crystals, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium bisulfate monohydrate, 97%, extra pure, reagent, crystals, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
10 kg
Packaging:
View all
Glass bottle
Catalog number 419430025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namesodium hydrate hydrogen sulfate
Molecular FormulaH3NaO5S
InChI KeyJXHZRQHZVYDRGX-UHFFFAOYSA-M
SMILESO.[Na+].OS([O-])(=O)=O
Molecular Weight (g/mol)138.07
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Adhering crystals
Titration with NaOH>=96.0 %
Chloride (Cl)=<20 ppm
Heavy metals (as Pb)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only