Sodium bisulfate monohydrate, 97%, extra pure, reagent, crystals
Sodium bisulfate monohydrate, 97%, extra pure, reagent, crystals
Sodium bisulfate monohydrate, 97%, extra pure, reagent, crystals
Sodium bisulfate monohydrate, 97%, extra pure, reagent, crystals
Thermo Scientific Chemicals

Sodium bisulfate monohydrate, 97%, extra pure, reagent, crystals

CAS: 10034-88-5 | H3NaO5S | 138.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 419435000
Price (USD)
125.00
Each
In stock
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
125.00
Each
Add to cart
Chemical Identifiers
CAS10034-88-5
IUPAC Namesodium hydrate hydrogen sulfate
Molecular FormulaH3NaO5S
InChI KeyJXHZRQHZVYDRGX-UHFFFAOYSA-M
SMILESO.[Na+].OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystals
Titration with NaOH>=96.0 %
Appearance (Color)White
Chloride (Cl)=<20 ppm
Heavy metals (as Pb)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection