Sodium bisulfite, ACS reagent, powder, Thermo Scientific Chemicals
Sodium bisulfite, ACS reagent, powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium bisulfite, ACS reagent, powder, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
100 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 419440010
Price (USD)/ Each
151.65
Online exclusive
178.00 
Save 26.35 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
151.65
Online exclusive
178.00 
Save 26.35 (15%)
Add to cart
Sodium bisulfite, ACS reagent, powder, Thermo Scientific Chemicals
Catalog number419440010
Price (USD)/ Each
151.65
Online exclusive
178.00 
Save 26.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namesodium sulfite
Molecular FormulaNaO3S
InChI KeyDWAQJAXMDSEUJJ-UHFFFAOYSA-L
SMILES[Na+].[O-]S([O-])=O
Molecular Weight (g/mol)103.05
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Iodimetric>=58.5 % (SO2)
Heavy metals (as Pb)=<10 ppm
Iron (Fe)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only