Chemical Identifiers
CAS1303-96-4
IUPAC Namedisodium decahydrate oxoboranyl ({oxido[(oxoboranyl)oxy]boranyl}oxy)boronate
Molecular FormulaB4H20Na2O17
InChI KeyOJPFLODMGUZMKD-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B(OB=O)OB([O-])OB=O
View more