Thermo Scientific Chemicals

Sodium tetraborate decahydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 419450025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium tetraborate decahydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 419450025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 419450025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1303-96-4
IUPAC Name
disodium decahydrate oxoboranyl ({oxido[(oxoboranyl)oxy]boranyl}oxy)boronate
Molecular Formula
B4H20Na2O17
InChI Key
OJPFLODMGUZMKD-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B(OB=O)OB([O-])OB=O
Appearance (Form)
Crystalline powder
Titration with NaOH
99.5 to 105.0 %
Heavy metals (as Pb)
=<10 ppm
Insoluble matter
=<0.005 %
pH
9.15 to 9.20 (0.01 M solution at 25.0°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text