Sodium carbonate monohydrate, 99.6%, ACS reagent
Sodium carbonate monohydrate, 99.6%, ACS reagent
Sodium carbonate monohydrate, 99.6%, ACS reagent
Sodium carbonate monohydrate, 99.6%, ACS reagent
Thermo Scientific Chemicals

Sodium carbonate monohydrate, 99.6%, ACS reagent

CAS: 5968-11-6 | CNa2O3·H2O | 124.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 419480010
Price (USD)
131.65
Online Exclusive
146.00
Save 14.35 (10%)
Each
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
131.65
Online Exclusive
146.00
Save 14.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS5968-11-6
IUPAC Namedisodium hydrate carbonate
Molecular FormulaCH2Na2O4
InChI KeyMQRJBSHKWOFOGF-UHFFFAOYSA-L
SMILESO.[Na+].[Na+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Heavy metals (as Pb)=<5 ppm
Insoluble matter=<0.01 %
Sulfur compounds (as SO4)=<0.004 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection