Thermo Scientific Chemicals

Sodium carbonate monohydrate, 99.6%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 419480051
5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium carbonate monohydrate, 99.6%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 419480051
5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 419480051
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5968-11-6
IUPAC Name
disodium hydrate carbonate
Molecular Formula
CH2Na2O4
InChI Key
MQRJBSHKWOFOGF-UHFFFAOYSA-L
SMILES
O.[Na+].[Na+].[O-]C([O-])=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Heavy metals (as Pb)
=<5 ppm
Insoluble matter
=<0.01 %
Sulfur compounds (as SO4)
=<0.004 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text