Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis
Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis
Thermo Scientific Chemicals

Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
1 kg
Catalog number 419530050
Price (USD)/ Each
-
Quantity:
5 kg
Request bulk or custom format
Chemical Identifiers
CAS151-21-3, 7757-82-6, 7647-14-5
SpecificationsSpecification SheetSpecification Sheet
UVat 260 nm A: =<0.05
UVat 280 nm A: =<0.04
Solubility(3 % in water) Clear colorless
Appearance (Color)White
Appearance (Form)Crystalline powder or flakes
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
RUO – Research Use Only