Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis
Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis
Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis
Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis
Thermo Scientific Chemicals

Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Catalog number 419531000
Price (USD)
71.65
Special offer
Online exclusive
Ends: 26-Sep-2025
88.40
Save 16.75 (19%)
Each
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
71.65
Special offer
Online exclusive
Ends: 26-Sep-2025
88.40
Save 16.75 (19%)
Each
Add to cart
Chemical Identifiers
CAS151-21-3
IUPAC Namesodium dodecyl sulfate
Molecular FormulaC12H25NaO4S
InChI KeyDBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 260 nm A: =<0.05
UVat 280 nm A: =<0.04
Solubility(3 % in water) Clear colorless
Appearance (Color)White
Appearance (Form)Crystalline powder or flakes
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link