Sodium hexafluoroantimonate, 98%, Thermo Scientific Chemicals
Sodium hexafluoroantimonate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium hexafluoroantimonate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 419541000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
Molecular FormulaF6NaSb
InChI KeyZJLVIRKMUXZEDU-UHFFFAOYSA-N
SMILES[F].[F].[F].[F].[F].[F].[Na+].[Sb]
Molecular Weight (g/mol)258.74
CAS16925-25-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Titration with NaOH97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only