Thermo Scientific Chemicals

Sodium hypochlorite, 4-6%, Thermo Scientific Chemicals

Catalog number: 419550100
10 L, Plastic drum, Each
Thermo Scientific Chemicals

Sodium hypochlorite, 4-6%, Thermo Scientific Chemicals

Catalog number: 419550100
10 L, Plastic drum, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 419550100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7681-52-9
IUPAC Name
sodium hypochlorite
Molecular Formula
ClNaO
InChI Key
SUKJFIGYRHOWBL-UHFFFAOYSA-N
SMILES
[Na+].[O-]Cl
Appearance (Color)
Clear yellow
Appearance (Form)
Solution
Titration Iodometric
4.0 to 6.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text