Sodium hypochlorite, 4-6%
Sodium hypochlorite, 4-6%
Sodium hypochlorite, 4-6%
Sodium hypochlorite, 4-6%
Thermo Scientific Chemicals

Sodium hypochlorite, 4-6%

CAS: 7681-52-9 | ClNaO | 74.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
25 mL
1 L
2.5 L
10 L
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 419552500
Price (USD)
52.00
Each
-
Add to cart
Quantity:
250 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
52.00
Each
Add to cart
Chemical Identifiers
CAS7681-52-9
IUPAC Namesodium hypochlorite
Molecular FormulaClNaO
InChI KeySUKJFIGYRHOWBL-UHFFFAOYSA-N
SMILES[Na+].[O-]Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Appearance (Form)Solution
Titration Iodometric4.0 to 6.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link