Sodium iodide, ACS reagent, anhydrous, Thermo Scientific Chemicals
Sodium iodide, ACS reagent, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium iodide, ACS reagent, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Catalog number 419570025
Price (USD)/ Each
858.65
Online exclusive
954.00 
Save 95.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)/ Each
858.65
Online exclusive
954.00 
Save 95.35 (10%)
Add to cart
Sodium iodide, ACS reagent, anhydrous, Thermo Scientific Chemicals
Catalog number419570025
Price (USD)/ Each
858.65
Online exclusive
954.00 
Save 95.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7681-82-5
IUPAC Namesodium iodide
Molecular FormulaINa
InChI KeyFVAUCKIRQBBSSJ-UHFFFAOYSA-M
SMILES[Na+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<0.005 %
Barium (Ba)=<0.002 %
Calcium (Ca)=<0.002 %
Magnesium (Mg)=<0.001 %
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only