Sodium metabisulfite, 97+%, ACS reagent, Thermo Scientific Chemicals
Sodium metabisulfite, 97+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium metabisulfite, 97+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
View all
Glass Bottle
Catalog number 419580050
Price (USD)/ Each
30.65
Online exclusive
35.50 
Save 4.85 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
30.65
Online exclusive
35.50 
Save 4.85 (14%)
Add to cart
Sodium metabisulfite, 97+%, ACS reagent, Thermo Scientific Chemicals
Catalog number419580050
Price (USD)/ Each
30.65
Online exclusive
35.50 
Save 4.85 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Namedisodium (sulfinatooxy)sulfinate
Molecular FormulaNa2O5S2
InChI KeyLDTLADDKFLAYJA-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)OS([O-])=O
Molecular Weight (g/mol)190.09
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Iodimetric>=97.0 %
Heavy metals (as Pb)=<10 ppm
Insoluble matter=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only