Sodium metasilicate nonahydrate, 44-47.5% total solids, Thermo Scientific Chemicals
Sodium metasilicate nonahydrate, 44-47.5% total solids, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium metasilicate nonahydrate, 44-47.5% total solids, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 419590250
Price (USD)/ Each
19.65
Online exclusive
21.30 
Save 1.65 (8%)
-
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
19.65
Online exclusive
21.30 
Save 1.65 (8%)
Add to cart
Sodium metasilicate nonahydrate, 44-47.5% total solids, Thermo Scientific Chemicals
Catalog number419590250
Price (USD)/ Each
19.65
Online exclusive
21.30 
Save 1.65 (8%)
-
Add to cart
Chemical Identifiers
CAS13517-24-3
IUPAC Namedisodium oxosilanebis(olate) nonahydrate
Molecular FormulaH18Na2O12Si
InChI KeyPHIQPXBZDGYJOG-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay have a yellow-pink hue
Assay44 to 47.5 % total solids
Ratio1.02 to 1.06 molar ratio SiO2/Na2O
Appearance (Color)White
Appearance (Form)Granular powder or granules
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only