Thermo Scientific Chemicals

Sodium periodate, 99.8+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 419615000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium periodate, 99.8+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 419615000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 419615000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-28-5
IUPAC Name
sodium periodate
Molecular Formula
INaO4
InChI Key
JQWHASGSAFIOCM-UHFFFAOYSA-M
SMILES
[Na+].[O-][I](=O)(=O)=O
Other halogens (as Cl)
=<0.02 %
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Titration Iodometric
99.8 to 100.3 % (on dried basis)
Manganese (Mn)
=<3 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text