Starch, Potato, Powder, Thermo Scientific Chemicals
Starch, Potato, Powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Starch, Potato, Powder, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Plastic bottle
Catalog number 419690025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS9005-25-8
IUPAC Name2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyGUBGYTABKSRVRQ-UHFFFAOYNA-N
SMILESOCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<20.5 % (1 g, 130°C, 90 min)
Arsenic (As)=<0.50 ppm
Cadmium (Cd)=<0.10 ppm
Mercury (Hg)=<0.03 ppm
Lead (Pb)=<0.50 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only