1x1 image pixel for data collection
Starch, extra pure, potato, powder
Starch, extra pure, potato, powder
Starch, extra pure, potato, powder
Starch, extra pure, potato, powder
Thermo Scientific Chemicals

Starch, extra pure, potato, powder

CAS: 9005-25-8 | C12H22O11 | 342.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 419695000
Price (USD)
80.65
Online Exclusive
89.50
Save 8.85 (10%)
Each
In stock
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
80.65
Online Exclusive
89.50
Save 8.85 (10%)
Each
Add to cart
Chemical Identifiers
CAS9005-25-8
IUPAC Name2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyGUBGYTABKSRVRQ-UHFFFAOYNA-N
SMILESOCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<20.5 % (1 g, 130°C, 90 min)
pH6 to 8 (50 % suspension in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection