Stoddard Solvent, Thermo Scientific Chemicals
Stoddard Solvent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Stoddard Solvent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 419730010
Price (USD)/ Each
125.65
Online exclusive
139.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
125.65
Online exclusive
139.00 
Save 13.35 (10%)
Add to cart
Stoddard Solvent, Thermo Scientific Chemicals
Catalog number419730010
Price (USD)/ Each
125.65
Online exclusive
139.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Name6-bromohexyl acetate
Molecular FormulaC8H15BrO2
InChI KeyIMSPZLCTPSXORJ-UHFFFAOYSA-N
SMILESCC(=O)OCCCCCCBr
Molecular Weight (g/mol)223.11
View more
SpecificationsSpecification SheetSpecification Sheet
Corrosiveness=<1 (Copper corrosion) (ASTM D 130)
Corrosiveness=<1 (Copper corrosion) (ASTM D 130)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only