Stoddard Solvent, Thermo Scientific Chemicals
Stoddard Solvent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Stoddard Solvent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number 419730050
Price (USD)/ Each
408.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
480.00 
Save 71.35 (15%)
-
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)/ Each
408.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
480.00 
Save 71.35 (15%)
Add to cart
Stoddard Solvent, Thermo Scientific Chemicals
Catalog number419730050
Price (USD)/ Each
408.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
480.00 
Save 71.35 (15%)
-
Add to cart
Chemical Identifiers
CAS8052-41-3
IUPAC Name6-bromohexyl acetate
Molecular FormulaC8H15BrO2
InChI KeyIMSPZLCTPSXORJ-UHFFFAOYSA-N
SMILESCC(=O)OCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<25 APHA (ASTM D 1209)
Bromine number=<5 g Br/100 g (ASTM D 1159)
Density(15.6°C) 754 to 820 kg/m³ (ASTM D 4052)
Appearance (Form)Clear liquid
Boiling point149°C to 213°C (ASTM D 86)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only