Thermo Scientific Chemicals

Tannic acid, ACS reagent, Thermo Scientific Chemicals

Catalog number: 419995000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Tannic acid, ACS reagent, Thermo Scientific Chemicals

Catalog number: 419995000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 419995000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1401-55-4
IUPAC Name
2,3-dihydroxy-5-[({3,4,5,6-tetrakis[3,4-dihydroxy-5-(3,4,5-trihydroxybenzoyloxy)benzoyloxy]oxan-2-yl}methoxy)carbonyl]phenyl 3,4,5-trihydroxybenzoate
Molecular Formula
C76H52O46
InChI Key
LRBQNJMCXXYXIU-UHFFFAOYNA-N
SMILES
OC1=CC(=CC(O)=C1O)C(=O)OC1=CC(=CC(O)=C1O)C(=O)OCC1OC(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C1OC(=O)C1=CC(O)=C(O)C(OC(=O)C2=CC(O)=C(O)C(O)=C2)=C1
Infrared spectrum
Conforms
Loss on drying
=<12.0 % (105°C)
Residue after ignition
=<0.5 %
Sugars, dextrin
Passes test
Zinc (Zn)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text