Magnesium sulfate heptahydrate, 99-100.5%, FCC, USP for parenteral use, GMP, Macron Fine Chemicals™
Magnesium sulfate heptahydrate, 99-100.5%, FCC, USP for parenteral use, GMP, Macron Fine Chemicals™
Avantor™

Magnesium sulfate heptahydrate, 99-100.5%, FCC, USP for parenteral use, GMP, Macron Fine Chemicals™

This product is GMP Manufactured and meets U.S.P Requirements.Weitere Informationen
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
2.5 kg
12 kg
Katalognummer 4200-20
Preis (EUR)
-
Angebot anfordern
Menge:
12 kg
Ask our AI about this Product
This product is GMP Manufactured and meets U.S.P Requirements.
Specifications
CAS10034-99-8
Chemischer Name oder MaterialMagnesium sulfate heptahydrate
FarbeFarblos
Schmelzpunkt1124°C
SummenformelMgSO4·7H2O
VerpackungMall STAKMOR Case
SynonymEpsom salts
GüteUSP
Molekulargewicht (g/mol)246.48
Reinheit (%)99-100.5%
Physikalische FormCrystalline
Menge12 kg
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Dokumente und Downloads

Zertifikate

0 Ergebnisse angezeigt, oben nach einem bestimmten Zertifikat suchen

Sicherheitsdatenblätter

Product Information

Geben Sie die Katalognummer, den Namen oder Link frei.

1x1 image pixel for data collection