Chemical Identifiers
CAS1176-74-5
IUPAC Namepotassium 2,6-dibromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)[2-(ethoxycarbonyl)phenyl]methyl]benzen-1-olate
Molecular FormulaC22H13Br4KO4
InChI KeyWCIQBKUTYDIBJC-UHFFFAOYSA-M
SMILES[K+].CCOC(=O)C1=CC=CC=C1C(C1=CC(Br)=C([O-])C(Br)=C1)=C1C=C(Br)C(=O)C(Br)=C1
View more