Tetrabutylammonium nitrate, 98%, Thermo Scientific Chemicals
Tetrabutylammonium nitrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrabutylammonium nitrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 420130250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nametetrabutylazanium nitrate
Molecular FormulaC16H36N2O3
InChI KeyQHOKENWFMZXSEU-UHFFFAOYSA-N
SMILES[O-][N+]([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol)304.48
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only