Thermo Scientific Chemicals

Tetrabutylammonium nitrate, 98%, Thermo Scientific Chemicals

Catalog number: 420131000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetrabutylammonium nitrate, 98%, Thermo Scientific Chemicals

Catalog number: 420131000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 420131000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1941-27-1
IUPAC Name
tetrabutylazanium nitrate
Molecular Formula
C16H36N2O3
InChI Key
QHOKENWFMZXSEU-UHFFFAOYSA-N
SMILES
[O-][N+]([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
Appearance (Color)
White
Infrared spectrum
Conforms
Titration with HClO4
>=97.5 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text