Thermo Scientific Chemicals

Tetrabutylammonium Perchlorate, Contains Max. 10% Water 98%, Thermo Scientific Chemicals

Catalog number: 420140250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetrabutylammonium Perchlorate, Contains Max. 10% Water 98%, Thermo Scientific Chemicals

Catalog number: 420140250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 420140250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1923-70-2
IUPAC Name
tetrabutylazanium perchlorate
Molecular Formula
C16H36ClNO4
InChI Key
KBLZDCFTQSIIOH-UHFFFAOYSA-M
SMILES
[O-][Cl](=O)(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
Appearance (Form)
Crystalline powder
Appearance (Color)
White
Infrared spectrum
Conforms
Titration
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent, Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text