Tetrabutylammonium Perchlorate, Contains Max. 10% Water 98%, Thermo Scientific Chemicals
Tetrabutylammonium Perchlorate, Contains Max. 10% Water 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrabutylammonium Perchlorate, Contains Max. 10% Water 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 420141000
Price (USD)/ Each
268.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
316.00 
Save 47.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
268.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
316.00 
Save 47.35 (15%)
Add to cart
Tetrabutylammonium Perchlorate, Contains Max. 10% Water 98%, Thermo Scientific Chemicals
Catalog number420141000
Price (USD)/ Each
268.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
316.00 
Save 47.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1923-70-2
IUPAC Nametetrabutylazanium perchlorate
Molecular FormulaC16H36ClNO4
InChI KeyKBLZDCFTQSIIOH-UHFFFAOYSA-M
SMILES[O-][Cl](=O)(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White
Infrared spectrumConforms
Titration>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent, Phase transfer catalyst
RUO – Research Use Only