Thermo Scientific Chemicals

3,3',4',5-Tetrachlorosalicylanilide, 99%, Thermo Scientific Chemicals

Catalog number: 420230250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3,3',4',5-Tetrachlorosalicylanilide, 99%, Thermo Scientific Chemicals

Catalog number: 420230250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 420230250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1154-59-2
IUPAC Name
3,5-dichloro-N-(3,4-dichlorophenyl)-2-hydroxybenzamide
Molecular Formula
C13H7Cl4NO2
InChI Key
SJQBHPJLLIJASD-UHFFFAOYSA-N
SMILES
OC1=C(Cl)C=C(Cl)C=C1C(=O)NC1=CC=C(Cl)C(Cl)=C1
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Appearance (Color)
White to beige
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text