Tetradecyl sodium sulfate, 95%, Thermo Scientific Chemicals
Tetradecyl sodium sulfate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetradecyl sodium sulfate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 420260050
Price (USD)/ Each
133.65
Online exclusive
148.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
133.65
Online exclusive
148.00 
Save 14.35 (10%)
Add to cart
Tetradecyl sodium sulfate, 95%, Thermo Scientific Chemicals
Catalog number420260050
Price (USD)/ Each
133.65
Online exclusive
148.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1191-50-0
IUPAC Namesodium tetradecyl sulfate
Molecular FormulaC14H29NaO4S
InChI KeyUPUIQOIQVMNQAP-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Assay>=94 %
Appearance (Color)White to off-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only