Thermo Scientific Chemicals

Tetradecyl sodium sulfate, 95%, Thermo Scientific Chemicals

Catalog number: 420260250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetradecyl sodium sulfate, 95%, Thermo Scientific Chemicals

Catalog number: 420260250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 420260250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1191-50-0
IUPAC Name
sodium tetradecyl sulfate
Molecular Formula
C14H29NaO4S
InChI Key
UPUIQOIQVMNQAP-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O
Appearance (Form)
Crystalline powder
Assay
>=94 %
Appearance (Color)
White to off-white
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text