Tetraethylammonium hydroxide, 25% in water, Thermo Scientific Chemicals
Tetraethylammonium hydroxide, 25% in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetraethylammonium hydroxide, 25% in water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 420291000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nametetraethylazanium hydroxide
Molecular FormulaC8H21NO
InChI KeyLRGJRHZIDJQFCL-UHFFFAOYSA-M
SMILES[OH-].CC[N+](CC)(CC)CC
Molecular Weight (g/mol)147.26
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear colorless to light yellow
Titration with HCl24 to 26 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only