Tetraethyleneglycol monomethyl ether, 98%, Thermo Scientific Chemicals
Tetraethyleneglycol monomethyl ether, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetraethyleneglycol monomethyl ether, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 420350250
Price (USD)/ Each
207.65
Online exclusive
230.00 
Save 22.35 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
207.65
Online exclusive
230.00 
Save 22.35 (10%)
Add to cart
Tetraethyleneglycol monomethyl ether, 98%, Thermo Scientific Chemicals
Catalog number420350250
Price (USD)/ Each
207.65
Online exclusive
230.00 
Save 22.35 (10%)
-
Add to cart
Chemical Identifiers
CAS23783-42-8
IUPAC Name2,5,8,11-tetraoxatridecan-13-ol
Molecular FormulaC9H20O5
InChI KeyZNYRFEPBTVGZDN-UHFFFAOYSA-N
SMILESCOCCOCCOCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4440 to 1.4460 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only