Tetraethyl orthosilicate, 98%, AcroSeal™, Thermo Scientific Chemicals
Tetraethyl orthosilicate, 98%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetraethyl orthosilicate, 98%, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 420360010
Price (USD)/ Each
73.65
Online exclusive
82.20 
Save 8.55 (10%)
-
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
73.65
Online exclusive
82.20 
Save 8.55 (10%)
Add to cart
Tetraethyl orthosilicate, 98%, AcroSeal™, Thermo Scientific Chemicals
Catalog number420360010
Price (USD)/ Each
73.65
Online exclusive
82.20 
Save 8.55 (10%)
-
Add to cart
Chemical Identifiers
CAS78-10-4
IUPAC Nametetraethyl silicate
Molecular FormulaC8H20O4Si
InChI KeyBOTDANWDWHJENH-UHFFFAOYSA-N
SMILESCCO[Si](OCC)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
GC>=97.5 %
Color scale=<20 APHA
Refractive index1.3820 to 1.3840 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only