Thermo Scientific Chemicals

Tetrahydrofurfuryl alcohol, 99+%, Thermo Scientific Chemicals

Catalog number: 420440010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Tetrahydrofurfuryl alcohol, 99+%, Thermo Scientific Chemicals

Catalog number: 420440010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 420440010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
97-99-4
IUPAC Name
(oxolan-2-yl)methanol
Molecular Formula
C5H10O2
InChI Key
BSYVTEYKTMYBMK-UHFFFAOYNA-N
SMILES
OCC1CCCO1
Appearance (Form)
Clear liquid
GC
=<0.1 % Furfuryl alcohol
Infrared spectrum
Conforms
Water
=<0.3 %
GC
=<1.6 % 1,2-Pentanediol

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text