Tetrahydrofurfuryl alcohol, 99+%, Thermo Scientific Chemicals
Tetrahydrofurfuryl alcohol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrahydrofurfuryl alcohol, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
Packaging:
Glass bottle
Catalog number 420440025
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
106.00 
Save 15.35 (14%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
106.00 
Save 15.35 (14%)
Add to cart
Tetrahydrofurfuryl alcohol, 99+%, Thermo Scientific Chemicals
Catalog number420440025
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
106.00 
Save 15.35 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Name(oxolan-2-yl)methanol
Molecular FormulaC5H10O2
InChI KeyBSYVTEYKTMYBMK-UHFFFAOYNA-N
SMILESOCC1CCCO1
Molecular Weight (g/mol)102.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC=<0.1 % Furfuryl alcohol
Infrared spectrumConforms
Water=<0.3 %
GC=<1.6 % 1,2-Pentanediol
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only