Thermo Scientific Chemicals

Tetramethylammonium Hydroxide, 10% in Water, Thermo Scientific Chemicals

Catalog number: 420510010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Tetramethylammonium Hydroxide, 10% in Water, Thermo Scientific Chemicals

Catalog number: 420510010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 420510010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
75-59-2, 7732-18-5
Appearance (Color)
Clear colorless
Titration with HCl
9.5 to 10.5 %
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text