Tetramethylammonium Hydroxide, 10% in Water, Thermo Scientific Chemicals
Tetramethylammonium Hydroxide, 10% in Water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetramethylammonium Hydroxide, 10% in Water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number 420515000
Price (USD)/ Each
97.65
Online exclusive
115.00 
Save 17.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
97.65
Online exclusive
115.00 
Save 17.35 (15%)
Add to cart
Tetramethylammonium Hydroxide, 10% in Water, Thermo Scientific Chemicals
Catalog number420515000
Price (USD)/ Each
97.65
Online exclusive
115.00 
Save 17.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametetramethylazanium hydroxide
Molecular FormulaC4H13NO
InChI KeyWGTYBPLFGIVFAS-UHFFFAOYSA-M
SMILES[OH-].C[N+](C)(C)C
Molecular Weight (g/mol)91.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Titration with HCl9.5 to 10.5 %
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only