Tetramethylammonium hydroxide pentahydrate, 98%, Thermo Scientific Chemicals
Tetramethylammonium hydroxide pentahydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetramethylammonium hydroxide pentahydrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number 420530250
Price (USD)/ Each
72.65
Online exclusive
80.70 
Save 8.05 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
72.65
Online exclusive
80.70 
Save 8.05 (10%)
Add to cart
Tetramethylammonium hydroxide pentahydrate, 98%, Thermo Scientific Chemicals
Catalog number420530250
Price (USD)/ Each
72.65
Online exclusive
80.70 
Save 8.05 (10%)
-
Add to cart
Chemical Identifiers
CAS10424-65-4
IUPAC Nametetramethylazanium pentahydrate hydroxide
Molecular FormulaC4H23NO6
InChI KeyMYXKPFMQWULLOH-UHFFFAOYSA-M
SMILESO.O.O.O.O.[OH-].C[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Melting point68°C to 73°C
Carbonate (CO3)=<7500 ppm
Impurity=<0.1 % Halides (as Cl)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only