Thermo Scientific Chemicals

Tetramethylammonium hydroxide pentahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 420531000
100 g, Glass Bottle, Each
Thermo Scientific Chemicals

Tetramethylammonium hydroxide pentahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 420531000
100 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 420531000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10424-65-4
IUPAC Name
tetramethylazanium pentahydrate hydroxide
Molecular Formula
C4H23NO6
InChI Key
MYXKPFMQWULLOH-UHFFFAOYSA-M
SMILES
O.O.O.O.O.[OH-].C[N+](C)(C)C
Appearance (Color)
White to off-white
Melting point
68°C to 73°C
Carbonate (CO3)
=<7500 ppm
Impurity
=<0.1 % Halides (as Cl)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text