N,N,N',N'-Tetramethylethylenediamine, 99%, for biochemistry, For electrophoresis
N,N,N',N'-Tetramethylethylenediamine, 99%, for biochemistry, For electrophoresis
Thermo Scientific Chemicals

N,N,N',N'-Tetramethylethylenediamine, 99%, for biochemistry, For electrophoresis

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 420580500
Price (USD)/ Each
89.65
Online exclusive
99.30 
Save 9.65 (10%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
89.65
Online exclusive
99.30 
Save 9.65 (10%)
Add to cart
N,N,N',N'-Tetramethylethylenediamine, 99%, for biochemistry, For electrophoresis
Catalog number420580500
Price (USD)/ Each
89.65
Online exclusive
99.30 
Save 9.65 (10%)
-
Add to cart
Chemical Identifiers
CAS110-18-9
IUPAC Name[2-(dimethylamino)ethyl]dimethylamine
Molecular FormulaC6H16N2
InChI KeyKWYHDKDOAIKMQN-UHFFFAOYSA-N
SMILESCN(C)CCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4165 to 1.4195 (20°C, 589 nm)
Appearance (Color)Clear colorless to slightly turbid yellow
Appearance (Form)Liquid
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only