N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%
Thermo Scientific Chemicals

N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 420600250
Price (USD)/ Each
179.65
Online exclusive
199.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
179.65
Online exclusive
199.00 
Save 19.35 (10%)
Add to cart
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%
Catalog number420600250
Price (USD)/ Each
179.65
Online exclusive
199.00 
Save 19.35 (10%)
-
Add to cart
Chemical Identifiers
CAS637-01-4
IUPAC Namedihydrogen N1,N1,N4,N4-tetramethylbenzene-1,4-diamine dichloride
Molecular FormulaC10H18Cl2N2
InChI KeyFBHKTSXMTASXFJ-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].CN(C)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to grey to light brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO498.0 to 102.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only